accessibility menu, dialog, popup

UserName

OHBrBr

CAS RN 2050-49-9

IUPAC Name: 2,4-dibromonaphthalen-1-ol
Synonyms: 2,4-dibromonaphthalen-1-ol
Molecular Weight (g/mol): 301.97
Molecular Formula: C10H6Br2O
InChi Key: PSGUDVJPEWTBRM-UHFFFAOYSA-N
SMILES: OC1=C2C=CC=CC2=C(Br)C=C1Br
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results