missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dibromo-1-fluorobenzene 97.0+%, TCI America™
$46.20 - $225.25
Chemical Identifiers
| CAS | 1435-53-6 |
|---|---|
| Molecular Formula | C6H3Br2F |
| Molecular Weight (g/mol) | 253.90 |
| MDL Number | MFCD00000283 |
| InChI Key | MKHDDTWHDFVYDQ-UHFFFAOYSA-N |
| Synonym | 2,4-dibromofluorobenzene, 1,3-dibromo-4-fluorobenzene, 1-fluoro-2,4-dibromobenzene, benzene, 2,4-dibromo-1-fluoro, pubchem3446, acmc-209cqz, 1,3-dibromo-5-fluoro, intermediates-zcf02097, ksc494g9j |
| PubChem CID | 15034 |
| IUPAC Name | 2,4-dibromo-1-fluorobenzene |
| SMILES | FC1=CC=C(Br)C=C1Br |
Chemical Identifiers
| 1435-53-6 | |
| 253.90 | |
| MKHDDTWHDFVYDQ-UHFFFAOYSA-N | |
| 15034 | |
| FC1=CC=C(Br)C=C1Br |
| C6H3Br2F | |
| MFCD00000283 | |
| 2,4-dibromofluorobenzene, 1,3-dibromo-4-fluorobenzene, 1-fluoro-2,4-dibromobenzene, benzene, 2,4-dibromo-1-fluoro, pubchem3446, acmc-209cqz, 1,3-dibromo-5-fluoro, intermediates-zcf02097, ksc494g9j | |
| 2,4-dibromo-1-fluorobenzene |
Specifications
| 1435-53-6 | |
| C6H3Br2F | |
| 5 g | |
| MKHDDTWHDFVYDQ-UHFFFAOYSA-N | |
| 2,4-dibromo-1-fluorobenzene | |
| 15034 | |
| ≥97.0% (GC) | |
| 2,4-Dibromo-1-fluorobenzene |
| 105°C | |
| MFCD00000283 | |
| 2,4-dibromofluorobenzene, 1,3-dibromo-4-fluorobenzene, 1-fluoro-2,4-dibromobenzene, benzene, 2,4-dibromo-1-fluoro, pubchem3446, acmc-209cqz, 1,3-dibromo-5-fluoro, intermediates-zcf02097, ksc494g9j | |
| FC1=CC=C(Br)C=C1Br | |
| 253.90 | |
| 253.90 | |
| Liquid |
Safety and Handling
TSCA : No