accessibility menu, dialog, popup

UserName

FBrBr

CAS RN 1435-53-6

IUPAC Name: 2,4-dibromo-1-fluorobenzene
Synonyms: 2,4-dibromofluorobenzene
Molecular Weight (g/mol): 253.9
Molecular Formula: C6H3Br2F
InChi Key: MKHDDTWHDFVYDQ-UHFFFAOYSA-N
SMILES: FC1=CC=C(Br)C=C1Br
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)