missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diamino-6-dimethylamino-1,3,5-triazine 98.0+%, TCI America™

Supplier:  TCI America D22365G

 View more versions of this product

Catalog No. D22365G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,4-Diamino-6-dimethylamino-1,3,5-triazine
White
MFCD00191338
N2,N2-Dimethylmelamine
CN(C)C1=NC(=NC(=N1)N)N
154.177
154.18
Crystalline Powder
1985-46-2
C5H10N6
5 g
IEFWDQQGFDLKFK-UHFFFAOYSA-N
2-N,2-N-dimethyl-1,3,5-triazine-2,4,6-triamine
16134
≥98.0% (HPLC,T)

Chemical Identifiers

1985-46-2
154.177
IEFWDQQGFDLKFK-UHFFFAOYSA-N
16134
CN(C)C1=NC(=NC(=N1)N)N
C5H10N6
MFCD00191338
N2,N2-Dimethylmelamine
2-N,2-N-dimethyl-1,3,5-triazine-2,4,6-triamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : OS0875000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions