missing translation for 'onlineSavingsMsg'
Learn More

2,4-Diamino-6-dimethylamino-1,3,5-triazine 98.0+%, TCI America™

Catalog No. p-7127183
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22361G 1 g
D22365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22361G Supplier TCI America Supplier No. D22361G
Only null left

Chemical Identifiers

CAS 1985-46-2
Molecular Formula C5H10N6
Molecular Weight (g/mol) 154.177
MDL Number MFCD00191338
InChI Key IEFWDQQGFDLKFK-UHFFFAOYSA-N
Synonym N2,N2-Dimethylmelamine
PubChem CID 16134
IUPAC Name 2-N,2-N-dimethyl-1,3,5-triazine-2,4,6-triamine
SMILES CN(C)C1=NC(=NC(=N1)N)N

Specifications

Color White
Quantity 1 g
Formula Weight 154.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Diamino-6-dimethylamino-1,3,5-triazine
RTECSNumber OS0875000
TSCA No