accessibility menu, dialog, popup

UserName

H3CNCH3NNH2NH2NN

CAS RN 1985-46-2

IUPAC Name: N2,N2-dimethyl-1,3,5-triazine-2,4,6-triamine
Synonyms: N2,N2-dimethyl-1,3,5-triazine-2,4,6-triamine
Molecular Weight (g/mol): 154.18
Molecular Formula: C5H10N6
InChi Key: IEFWDQQGFDLKFK-UHFFFAOYSA-N
SMILES: CN(C)C1=NC(N)=NC(N)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results