missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Chlorophenoxy)-6-fluorobenzaldehyde, 98%

Catalog No. p-7033695
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50847MD 250 mg
AAH5084703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50847MD Supplier Thermo Scientific Chemicals Supplier No. H50847MD
Only null left

CAS: 902836-82-2 | C13H8ClFO2 | 250.653 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 902836-82-2
Molecular Formula C13H8ClFO2
Molecular Weight (g/mol) 250.653
MDL Number MFCD08061024
InChI Key JEPXYNGAXLVUMW-UHFFFAOYSA-N
Synonym 2-4-chlorophenoxy-6-fluorobenzaldehyde, 2-4-chloro-phenoxy-6-fluoro-benzaldehyde, acmc-20aopr
PubChem CID 42553314
IUPAC Name 2-(4-chlorophenoxy)-6-fluorobenzaldehyde
SMILES C1=CC(=C(C(=C1)F)C=O)OC2=CC=C(C=C2)Cl

Specifications

Quantity 250 mg
Formula Weight 250.66
Percent Purity 98%
Chemical Name or Material 2-(4-Chlorophenoxy)-6-fluorobenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only