missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Bromophenyl)ethyl Alcohol 97.0+%, TCI America™
Supplier: TCI America B191825G
Specifications
| 2-(4-Bromophenyl)ethyl Alcohol | |
| Yellow | |
| C8H9BrO | |
| 25 g | |
| PMOSJSPFNDUAFY-UHFFFAOYSA-N | |
| 2-(4-bromophenyl)ethanol | |
| 72851 | |
| ≥97.0% (GC) |
| 4654-39-1 | |
| 150°C | |
| MFCD00002897 | |
| 2-4-bromophenyl ethanol, 4-bromophenethyl alcohol, p-bromophenethyl alcohol, benzeneethanol, 4-bromo, 2-4-bromophenyl ethan-1-ol, 2-4-bromo-phenyl-ethanol, 2-4-bromophenyl ethyl alcohol, 2-p-bromophenyl ethanol, phenethyl alcohol, p-bromo, 4-bromobenzene ethanol | |
| C1=CC(=CC=C1CCO)Br | |
| 201.063 | |
| 201.06 | |
| Liquid |
Chemical Identifiers
| 4654-39-1 | |
| 201.063 | |
| PMOSJSPFNDUAFY-UHFFFAOYSA-N | |
| 72851 | |
| C1=CC(=CC=C1CCO)Br |
| C8H9BrO | |
| MFCD00002897 | |
| 2-4-bromophenyl ethanol, 4-bromophenethyl alcohol, p-bromophenethyl alcohol, benzeneethanol, 4-bromo, 2-4-bromophenyl ethan-1-ol, 2-4-bromo-phenyl-ethanol, 2-4-bromophenyl ethyl alcohol, 2-p-bromophenyl ethanol, phenethyl alcohol, p-bromo, 4-bromobenzene ethanol | |
| 2-(4-bromophenyl)ethanol |
Safety and Handling
TSCA : No