missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)ethyl Alcohol 97.0+%, TCI America™

Catalog No. p-7119353
Change view
Click to view available options
Quantity:
25 g
5 g
Catalog No. Quantity
B19185G 5 g
B191825G 25 g
2 options

Catalog No. B19185G

Supplier: TCI America B19185G

Only null left

Chemical Identifiers

CAS 4654-39-1
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD00002897
InChI Key PMOSJSPFNDUAFY-UHFFFAOYSA-N
Synonym 2-4-bromophenyl ethanol, 4-bromophenethyl alcohol, p-bromophenethyl alcohol, benzeneethanol, 4-bromo, 2-4-bromophenyl ethan-1-ol, 2-4-bromo-phenyl-ethanol, 2-4-bromophenyl ethyl alcohol, 2-p-bromophenyl ethanol, phenethyl alcohol, p-bromo, 4-bromobenzene ethanol
PubChem CID 72851
IUPAC Name 2-(4-bromophenyl)ethanol
SMILES C1=CC(=CC=C1CCO)Br

Specifications

Color Yellow
Boiling Point 150°C
Quantity 5 g
Formula Weight 201.06
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(4-Bromophenyl)ethyl Alcohol