missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(4-Bromophenyl)benzoxazole 98.0+%, TCI America™
Supplier: TCI America B50175G
Specifications
| 2-(4-Bromophenyl)benzoxazole | |
| 161°C | |
| C13H8BrNO | |
| 5 g | |
| BrC1=CC=C(C=C1)C1=NC2=CC=CC=C2O1 | |
| 274.12 | |
| 274.12 | |
| Crystalline Powder |
| 3164-13-4 | |
| White-Yellow | |
| MFCD00168898 | |
| RBVHJNZMSBQFDK-UHFFFAOYSA-N | |
| 2-(4-bromophenyl)-1,3-benzoxazole | |
| 12846715 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 3164-13-4 | |
| 274.12 | |
| RBVHJNZMSBQFDK-UHFFFAOYSA-N | |
| 2-(4-bromophenyl)-1,3-benzoxazole |
| C13H8BrNO | |
| MFCD00168898 | |
| 12846715 | |
| BrC1=CC=C(C=C1)C1=NC2=CC=CC=C2O1 |
Safety and Handling
TSCA : No