missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)benzoxazole 98.0+%, TCI America™

Catalog No. p-7123900
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50171G 1 g
B50175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50171G Supplier TCI America Supplier No. B50171G
Only null left

Chemical Identifiers

CAS 3164-13-4
Molecular Formula C13H8BrNO
Molecular Weight (g/mol) 274.12
MDL Number MFCD00168898
InChI Key RBVHJNZMSBQFDK-UHFFFAOYSA-N
PubChem CID 12846715
IUPAC Name 2-(4-bromophenyl)-1,3-benzoxazole
SMILES BrC1=CC=C(C=C1)C1=NC2=CC=CC=C2O1

Specifications

Melting Point 161°C
Color White-Yellow
Quantity 1 g
Formula Weight 274.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Bromophenyl)benzoxazole