accessibility menu, dialog, popup

UserName

NOBr

CAS RN 3164-13-4

IUPAC Name: 2-(4-bromophenyl)-1,3-benzoxazole
Synonyms: 2-(4-bromophenyl)-1,3-benzoxazole
Molecular Weight (g/mol): 274.12
Molecular Formula: C13H8BrNO
InChi Key: RBVHJNZMSBQFDK-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=NC2=CC=CC=C2O1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results