missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98.0+%, TCI America™

Catalog No. B47711G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47711G 1 g
B47715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47711G Supplier TCI America Supplier No. B47711G
Only null left

Chemical Identifiers

CAS 68716-49-4
Molecular Formula C12H16BBrO2
Molecular Weight (g/mol) 282.97
MDL Number MFCD11110553
InChI Key AZCNDGAXOZWQPV-UHFFFAOYSA-N
Synonym 1-Bromo-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene, 4-Bromophenylboronic Acid Pinacol Ester
PubChem CID 10755376
IUPAC Name 2-(4-bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CC=C(Br)C=C1

Specifications

Melting Point 71°C
Color White
Quantity 1 g
Formula Weight 282.97
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane