accessibility menu, dialog, popup

UserName

CAS RN 68716-49-4

BOOH3CCH3H3CH3CBr

CAS RN 68716-49-4

IUPAC Name: 2-(4-bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(4-bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 282.97
Molecular Formula: C12H16BBrO2
InChi Key: AZCNDGAXOZWQPV-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results