missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine 98.0+%, TCI America™

Catalog No. B36555G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36555G 5 g
B36551G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36555G Supplier TCI America Supplier No. B36555G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 927384-44-9
Molecular Formula C16H12BBrN2
Molecular Weight (g/mol) 323.00
MDL Number MFCD16038141
InChI Key NKGYQJVDGSWXFR-UHFFFAOYSA-N
Synonym 4-Bromo-1-(2,3-dihydro-1H-naphtho[1,8-de]-1,3,2-diazaborinyl)benzene, 2-(4-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine
PubChem CID 53384413
IUPAC Name 3-(4-bromophenyl)-2,4-diaza-3-boratricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene
SMILES BrC1=CC=C(C=C1)B1NC2=CC=CC3=CC=CC(N1)=C23

Specifications

Quantity 5 g
Formula Weight 323.00
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-(4-Bromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborine
TSCA No
Recommended Storage Refrigerator