accessibility menu, dialog, popup

UserName

CAS RN 927384-44-9

IUPAC Name: 3-(4-bromophenyl)-2,4-diaza-3-boratricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene
Synonyms: 3-(4-bromophenyl)-2,4-diaza-3-boratricyclo[7.3.1.0⁵,¹³]trideca-1(13),5,7,9,11-pentaene
Molecular Weight (g/mol): 323
Molecular Formula: C16H12BBrN2
InChi Key: NKGYQJVDGSWXFR-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)B1NC2=CC=CC3=CC=CC(N1)=C23
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results