missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Bromophenoxy)pyridine, ≥95%, Thermo Scientific™

Catalog No. CC48910CB
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC48910CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CC48910CB Supplier Thermo Scientific Chemicals Supplier No. CC48910CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4783-82-8
Molecular Formula C11H8BrNO
Molecular Weight (g/mol) 250.10
MDL Number MFCD08435900
InChI Key FOKJQGKEVMZAJE-UHFFFAOYSA-N
Synonym 2-4-bromophenoxy pyridine, pyridine,2-4-bromophenoxy, 2-4-bromo-phenoxy pyridine, 2-4-bromo-phenoxy-pyridine, pyridine, 2-4-bromophenoxy, 4-bromo-1-2-pyridyloxy benzene
PubChem CID 23201935
IUPAC Name 2-(4-bromophenoxy)pyridine
SMILES BrC1=CC=C(OC2=CC=CC=N2)C=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 250.09
Percent Purity 95%
Chemical Name or Material 2-(4-Bromophenoxy)pyridine
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.