Learn More
2,4-Bis(trifluoromethyl)benzyl bromide, 97%
CAS: 140690-56-8 | C9H5BrF6 | 307.033 g/mol
$122.52 - $487.22
Chemical Identifiers
| CAS | 140690-56-8 |
|---|---|
| Molecular Formula | C9H5BrF6 |
| Molecular Weight (g/mol) | 307.033 |
| MDL Number | MFCD00010306 |
| InChI Key | SWFFFUJOWAJJCH-UHFFFAOYSA-N |
| Synonym | 2,4-bis trifluoromethyl benzyl bromide, 1-bromomethyl-2,4-bis trifluoromethyl benzene, 2,4-bis trifluoromethyl benzylbromide, benzene, 1-bromomethyl-2,4-bis trifluoromethyl, 2,4-bis trifluoromethyl-1-bromomethyl benzene, pubchem4879, acmc-1c0tm, 2,4-bis-trifluoromethyl-benzylbromide, 2,4-bis-trifluoromethyl benzyl bromide, 2,4-bis-trifluoromethyl-benzyl bromide |
| PubChem CID | 518871 |
| IUPAC Name | 1-(bromomethyl)-2,4-bis(trifluoromethyl)benzene |
| SMILES | C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)CBr |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2391003
|
Thermo Scientific Chemicals
B2391003 |
1 g |
Each for $122.52
|
|
|||||
|
AAB2391006
|
Thermo Scientific Chemicals
B2391006 |
5 g |
Each for $487.22
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 140690-56-8 | |
| 307.033 | |
| SWFFFUJOWAJJCH-UHFFFAOYSA-N | |
| 518871 | |
| C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)CBr |
| C9H5BrF6 | |
| MFCD00010306 | |
| 2,4-bis trifluoromethyl benzyl bromide, 1-bromomethyl-2,4-bis trifluoromethyl benzene, 2,4-bis trifluoromethyl benzylbromide, benzene, 1-bromomethyl-2,4-bis trifluoromethyl, 2,4-bis trifluoromethyl-1-bromomethyl benzene, pubchem4879, acmc-1c0tm, 2,4-bis-trifluoromethyl-benzylbromide, 2,4-bis-trifluoromethyl benzyl bromide, 2,4-bis-trifluoromethyl-benzyl bromide | |
| 1-(bromomethyl)-2,4-bis(trifluoromethyl)benzene |
Specifications
| 140690-56-8 | |
| 112°C (0.2 mmHg) | |
| C9H5BrF6 | |
| MFCD00010306 | |
| UN3265 | |
| 2,4-bis trifluoromethyl benzyl bromide, 1-bromomethyl-2,4-bis trifluoromethyl benzene, 2,4-bis trifluoromethyl benzylbromide, benzene, 1-bromomethyl-2,4-bis trifluoromethyl, 2,4-bis trifluoromethyl-1-bromomethyl benzene, pubchem4879, acmc-1c0tm, 2,4-bis-trifluoromethyl-benzylbromide, 2,4-bis-trifluoromethyl benzyl bromide, 2,4-bis-trifluoromethyl-benzyl bromide | |
| C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)CBr | |
| 307.033 | |
| 307.04 | |
| 2,4-Bis(trifluoromethyl)benzyl bromide |
| 1.637 | |
| 92°C (197°F) | |
| 1.4511 | |
| 1 g | |
| 7995492 | |
| SWFFFUJOWAJJCH-UHFFFAOYSA-N | |
| 1-(bromomethyl)-2,4-bis(trifluoromethyl)benzene | |
| 518871 | |
| 97% |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only