CAS RN 140690-56-8
IUPAC Name:
1-(bromomethyl)-2,4-bis(trifluoromethyl)benzene
Synonyms:
1-(bromomethyl)-2,4-bis(trifluoromethyl)benzene
Molecular Weight (g/mol):
307.03
Molecular Formula:
C9H5BrF6
InChi Key:
SWFFFUJOWAJJCH-UHFFFAOYSA-N
SMILES:
FC(F)(F)C1=CC=C(CBr)C(=C1)C(F)(F)F
1
–
1
of
1
results
1
Promotions Available
2,4-Bis(trifluoromethyl)benzyl bromide, 97%
2,4-Bis(trifluoromethyl)benzyl bromide, 97%
CAS: 140690-56-8 Molecular Formula: C9H5BrF6 Molecular Weight (g/mol): 307.033 MDL Number: MFCD00010306 InChI Key: SWFFFUJOWAJJCH-UHFFFAOYSA-N Synonym: 2,4-bis trifluoromethyl benzyl bromide,1-bromomethyl-2,4-bis trifluoromethyl benzene,2,4-bis trifluoromethyl benzylbromide,benzene, 1-bromomethyl-2,4-bis trifluoromethyl,2,4-bis trifluoromethyl-1-bromomethyl benzene,pubchem4879,acmc-1c0tm,2,4-bis-trifluoromethyl-benzylbromide,2,4-bis-trifluoromethyl benzyl bromide,2,4-bis-trifluoromethyl-benzyl bromide PubChem CID: 518871 IUPAC Name: 1-(bromomethyl)-2,4-bis(trifluoromethyl)benzene SMILES: C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)CBr