missing translation for 'onlineSavingsMsg'
Learn More

2,4-Bis(trifluoromethyl)benzyl bromide, 97%

Catalog No. AAB2391003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2391003 1 g
AAB2391006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2391003 Supplier Thermo Scientific Chemicals Supplier No. B2391003

CAS: 140690-56-8 | C9H5BrF6 | 307.033 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 140690-56-8
Molecular Formula C9H5BrF6
Molecular Weight (g/mol) 307.033
MDL Number MFCD00010306
InChI Key SWFFFUJOWAJJCH-UHFFFAOYSA-N
Synonym 2,4-bis trifluoromethyl benzyl bromide, 1-bromomethyl-2,4-bis trifluoromethyl benzene, 2,4-bis trifluoromethyl benzylbromide, benzene, 1-bromomethyl-2,4-bis trifluoromethyl, 2,4-bis trifluoromethyl-1-bromomethyl benzene, pubchem4879, acmc-1c0tm, 2,4-bis-trifluoromethyl-benzylbromide, 2,4-bis-trifluoromethyl benzyl bromide, 2,4-bis-trifluoromethyl-benzyl bromide
PubChem CID 518871
IUPAC Name 1-(bromomethyl)-2,4-bis(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)C(F)(F)F)CBr

Specifications

Density 1.637
Boiling Point 112°C (0.2 mmHg)
Flash Point 92°C (197°F)
Refractive Index 1.4511
Quantity 1 g
UN Number UN3265
Beilstein 7995492
Formula Weight 307.04
Percent Purity 97%
Chemical Name or Material 2,4-Bis(trifluoromethyl)benzyl bromide
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only