missing translation for 'onlineSavingsMsg'
Learn More

2,4-Bis(chloromethyl)mesitylene, 98%

Catalog No. AAA1383809
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1383809 10 g
AAA1383818 50 g
AAA1383830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1383809 Supplier Thermo Scientific Chemicals Supplier No. A1383809

CAS: 1585-17-7 | C11H14Cl2 | 217.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1585-17-7
Molecular Formula C11H14Cl2
Molecular Weight (g/mol) 217.13
MDL Number MFCD00013681
InChI Key JMNDJDCLOQRCJV-UHFFFAOYSA-N
Synonym 2,4-bis chloromethyl-1,3,5-trimethylbenzene, 2,4-bis chloromethyl mesitylene, benzene, 2,4-bis chloromethyl-1,3,5-trimethyl, 2,4-di chloromethyl-1,3,5-trimethylbenzene, 2,4-bis chloromethylmesitylene, acmc-209dhz, bis-chloromethyl mesitylene, bis-chloromethyl-mesitylene, jmndjdcloqrcjv-uhfffaoysa, 2,4-bis-chloromethyl mesitylene
PubChem CID 137095
IUPAC Name 2,4-bis(chloromethyl)-1,3,5-trimethylbenzene
SMILES CC1=CC(C)=C(CCl)C(C)=C1CCl

Specifications

Melting Point 104°C to 107°C
Boiling Point 180°C (20 mmHg)
Quantity 10 g
UN Number UN3261
Beilstein 2520798
Formula Weight 217.14
Percent Purity 98%
Chemical Name or Material 2,4-Bis(chloromethyl)mesitylene
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only