accessibility menu, dialog, popup

UserName

CH3CH3ClH3CCl

CAS RN 1585-17-7

IUPAC Name: 2,4-bis(chloromethyl)-1,3,5-trimethylbenzene
Synonyms: 2,4-bis(chloromethyl)-1,3,5-trimethylbenzene
Molecular Weight (g/mol): 217.13
Molecular Formula: C11H14Cl2
InChi Key: JMNDJDCLOQRCJV-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(CCl)C(C)=C1CCl
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)