missing translation for 'onlineSavingsMsg'
Learn More

2,4-Bis(chloromethyl)-1,3,5-trimethylbenzene 98.0+%, TCI America™

Catalog No. B199425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B199425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B199425G Supplier TCI America Supplier No. B199425G

Chemical Identifiers

CAS 1585-17-7
Molecular Formula C11H14Cl2
Molecular Weight (g/mol) 217.13
MDL Number MFCD00013681
InChI Key JMNDJDCLOQRCJV-UHFFFAOYSA-N
Synonym 2,4-bis chloromethyl-1,3,5-trimethylbenzene, 2,4-bis chloromethyl mesitylene, benzene, 2,4-bis chloromethyl-1,3,5-trimethyl, 2,4-di chloromethyl-1,3,5-trimethylbenzene, 2,4-bis chloromethylmesitylene, acmc-209dhz, bis-chloromethyl mesitylene, bis-chloromethyl-mesitylene, jmndjdcloqrcjv-uhfffaoysa, 2,4-bis-chloromethyl mesitylene
PubChem CID 137095
IUPAC Name 2,4-bis(chloromethyl)-1,3,5-trimethylbenzene
SMILES CC1=CC(C)=C(CCl)C(C)=C1CCl

Specifications

Melting Point 107°C
Color White-Yellow
Boiling Point 180°C
Quantity 25 g
UN Number 3261
Formula Weight 217.13
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Bis(chloromethyl)-1,3,5-trimethylbenzene