missing translation for 'onlineSavingsMsg'
Learn More

2,4-Bis[8-hydroxy-1,1,7,7-tetramethyljulolidin-9-yl]squaraine 98.0+%, TCI America™

Catalog No. p-7123667
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46491G 1 g
B46495G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46491G Supplier TCI America Supplier No. B46491G
Only null left

Chemical Identifiers

CAS 358727-55-6
Molecular Formula C36H44N2O4-2
Molecular Weight (g/mol) 568.758
InChI Key ZVJJZBXHRPLGNI-UHFFFAOYSA-L
Synonym 2,4-Bis[8-hydroxy-1,1,7,7-tetramethyl-1,2,3,5,6,7-hexahydropyrido[3,2,1-ij]quinoline-9-yl]squaraine
PubChem CID 91972175
SMILES CC1(CCN2CCC(C3=C(C(=CC1=C32)C4=C(C(=C4O)C5=CC6=C7C(=C5[O-])C(CCN7CCC6(C)C)(C)C)O)[O-])(C)C)C

Specifications

Color Yellow
Quantity 1 g
Formula Weight 568.76
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 2,4-Bis[8-hydroxy-1,1,7,7-tetramethyljulolidin-9-yl]squaraine