missing translation for 'onlineSavingsMsg'
Learn More

2-(4-Benzyloxyphenyl)ethanol, 98+%

Catalog No. AAL1921706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1921706 5 g
AAL1921714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1921706 Supplier Thermo Scientific Chemicals Supplier No. L1921706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 61439-59-6 | C15H16O2 | 228.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61439-59-6
Molecular Formula C15H16O2
Molecular Weight (g/mol) 228.29
MDL Number MFCD00017532
InChI Key JCUJAHLWCDISCC-UHFFFAOYSA-N
Synonym 2-4-benzyloxyphenyl ethanol, 2-4-benzyloxy phenyl ethanol, p-benzyloxy phenethyl alcohol, 4-benzyloxyphenylethyl alcohol, 4-benzyloxyphenethyl alcohol, 4-benzyloxy phenethylalcohol, benzeneethanol, 4-phenylmethoxy, 2-p-benzyloxyphenyl ethanol, 4-benzyloxy phenethyl alcohol
PubChem CID 3017260
IUPAC Name 2-(4-phenylmethoxyphenyl)ethanol
SMILES OCCC1=CC=C(OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 85°C to 87°C
Quantity 5 g
Beilstein 2054624
Formula Weight 228.29
Percent Purity ≥98%
Chemical Name or Material 2-(4-Benzyloxyphenyl)ethanol
EINECSNumber 262-795-2
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only