missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tris(4-fluorophenyl)boroxin 98.0+%, TCI America™

Catalog No. T181410G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T181410G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T181410G Supplier TCI America Supplier No. T181410G

Chemical Identifiers

CAS 448-59-9
Molecular Formula C18H12B3F3O3
Molecular Weight (g/mol) 365.716
MDL Number MFCD02093496
InChI Key AHYNJLHYJZXUSD-UHFFFAOYSA-N
Synonym 4-Fluorophenylboronic Anhydride, Tris(4-fluorophenyl)cyclotriboroxane
PubChem CID 2769359
IUPAC Name 2,4,6-tris(4-fluorophenyl)-1,3,5,2,4,6-trioxatriborinane
SMILES B1(OB(OB(O1)C2=CC=C(C=C2)F)C3=CC=C(C=C3)F)C4=CC=C(C=C4)F

Specifications

Color White
Quantity 10 g
Formula Weight 365.72
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tris(4-fluorophenyl)boroxin