accessibility menu, dialog, popup

UserName

CAS RN 448-59-9

IUPAC Name: tris(4-fluorophenyl)-1,3,5,2,4,6-trioxatriborinane
Synonyms: tris(4-fluorophenyl)-1,3,5,2,4,6-trioxatriborinane
Molecular Weight (g/mol): 365.72
Molecular Formula: C18H12B3F3O3
InChi Key: AHYNJLHYJZXUSD-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1)B1OB(OB(O1)C1=CC=C(F)C=C1)C1=CC=C(F)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results