missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Tris(3',5'-di-tert-butyl-4'-hydroxybenzyl)mesitylene 95.0+%, TCI America™

Catalog No. p-7136496
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T091625G 25 g
T0916100G 100 g
T0916500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T091625G Supplier TCI America Supplier No. T091625G
Only null left

Chemical Identifiers

CAS 1709-70-2
Molecular Formula C54H78O3
Molecular Weight (g/mol) 775.215
MDL Number MFCD00026284
InChI Key VSAWBBYYMBQKIK-UHFFFAOYSA-N
PubChem CID 74370
IUPAC Name 4-[[3,5-bis[(3,5-ditert-butyl-4-hydroxyphenyl)methyl]-2,4,6-trimethylphenyl]methyl]-2,6-ditert-butylphenol
SMILES CC1=C(C(=C(C(=C1CC2=CC(=C(C(=C2)C(C)(C)C)O)C(C)(C)C)C)CC3=CC(=C(C(=C3)C(C)(C)C)O)C(C)(C)C)C)CC4=CC(=C(C(=C4)C(C)(C)C)O)C(C)(C)C

Specifications

Melting Point 243°C
Color White
Quantity 25 g
Formula Weight 775.22
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Tris(3′,5′-di-tert-butyl-4′-hydroxybenzyl)mesitylene
EINECSNumber (4)-0191
RTECSNumber DC3750000
TSCA Yes