accessibility menu, dialog, popup

UserName

CAS RN 1709-70-2

CH3OHCH3H3CCH3CH3CH3H3CCH3HOH3CCH3H3CCH3H3CCH3H3COHCH3CH3H3CH3CCH3H3C

CAS RN 1709-70-2

IUPAC Name: 4-({3,5-bis[(3,5-di-tert-butyl-4-hydroxyphenyl)methyl]-2,4,6-trimethylphenyl}methyl)-2,6-di-tert-butylphenol
Synonyms: ethanox 330 tris-bht mesitylene
Molecular Weight (g/mol): 775.22
Molecular Formula: C54H78O3
InChi Key: VSAWBBYYMBQKIK-UHFFFAOYSA-N
SMILES: CC1=C(CC2=CC(=C(O)C(=C2)C(C)(C)C)C(C)(C)C)C(C)=C(CC2=CC(=C(O)C(=C2)C(C)(C)C)C(C)(C)C)C(C)=C1CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results