missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Tris(3',5'-di-tert-butyl-4'-hydroxybenzyl)mesitylene 95.0+%, TCI America™
Specifications
Specifications
| Melting Point | 243°C |
| Color | White |
| Quantity | 500 g |
| Formula Weight | 775.22 |
| Percent Purity | ≥95.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,4,6-Tris(3′,5′-di-tert-butyl-4′-hydroxybenzyl)mesitylene |
Safety and Handling
| EINECSNumber | (4)-0191 |
| RTECSNumber | DC3750000 |
| TSCA | Yes |