missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Triphenoxy-1,3,5-triazine, 96%, Thermo Scientific Chemicals

Catalog No. AAH3357218
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3357218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3357218 Supplier Thermo Scientific Chemicals Supplier No. H3357218
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1919-48-8 | Molecular Formula: C21H15N3O3 | Molecular Weight: 357.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1919-48-8
Molecular Formula C21H15N3O3
Molecular Weight (g/mol) 357.37
MDL Number MFCD00006048
InChI Key IYDYVVVAQKFGBS-UHFFFAOYSA-N
PubChem CID 15973
IUPAC Name 2,4,6-triphenoxy-1,3,5-triazine
SMILES O(C1=CC=CC=C1)C1=NC(OC2=CC=CC=C2)=NC(OC2=CC=CC=C2)=N1

Specifications

Melting Point 235°C to 236°C
Quantity 50 g
Synonym triphenyl cyanurate, triphenoxy-s-triazine, 1,3,5-triazine, 2,4,6-triphenoxy, 2,4,6-triphenoxy-s-triazine, s-triazine, 2,4,6-triphenoxy, triphenoxy-1,3,5-triazine, 1,3,5-triazine,2,4,6-triphenoxy, triazine der., maybridge4_003089, 2,6-triphenoxy-s-triazine
Formula Weight 357.36
Percent Purity 96%
Chemical Name or Material 2,4,6-Triphenoxy-1,3,5-triazine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 217-644-5
RTECSNumber XZ2030000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only