accessibility menu, dialog, popup

UserName

CAS RN 1919-48-8

ONONON

CAS RN 1919-48-8

IUPAC Name: triphenoxy-1,3,5-triazine
Synonyms: triphenoxy-1,3,5-triazine
Molecular Weight (g/mol): 357.37
Molecular Formula: C21H15N3O3
InChi Key: IYDYVVVAQKFGBS-UHFFFAOYSA-N
SMILES: O(C1=CC=CC=C1)C1=NC(OC2=CC=CC=C2)=NC(OC2=CC=CC=C2)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results