missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylthiophenol, 97%

Catalog No. AAB2479803
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2479803 1 g
AAB2479806 5 g
AAB2479814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2479803 Supplier Thermo Scientific Chemicals Supplier No. B2479803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1541-10-2 | C9H12S | 152.255 g/mol

2,4,6-Trimethylthiophenol, is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylthiophenol, is used as pharmaceutical intermediate.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1541-10-2
Molecular Formula C9H12S
Molecular Weight (g/mol) 152.255
MDL Number MFCD00218475
InChI Key PKANQZUPJCMBAK-UHFFFAOYSA-N
Synonym 2,4,6-trimethylthiophenol, 2,4,6-trimethylbenzene-1-thiol, benzenethiol,2,4,6-trimethyl, 2-mercaptomesitylene, 2-sulphanylmesitylene, acmc-20aoem, 2,4,6-trimethyl-benzene-1-thiol
PubChem CID 284449
IUPAC Name 2,4,6-trimethylbenzenethiol
SMILES CC1=CC(=C(C(=C1)C)S)C

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 152.25
Percent Purity 97%
Chemical Name or Material 2,4,6-Trimethylthiophenol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only