accessibility menu, dialog, popup

UserName

CAS RN 1541-10-2

IUPAC Name: (2,4,6-trimethylphenyl)sulfanide
Synonyms: (2,4,6-trimethylphenyl)sulfanide
Molecular Weight (g/mol): 151.25
Molecular Formula: C9H11S
InChi Key: PKANQZUPJCMBAK-UHFFFAOYSA-M
SMILES: CC1=CC(C)=C([S-])C(C)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)