missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylstyrene (stabilized with TBC) 97.0+%, TCI America™

Catalog No. T04931ML
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T04931ML 1 mL
T04935ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T04931ML Supplier TCI America Supplier No. T04931ML

Chemical Identifiers

CAS 769-25-5
Molecular Formula C11H14
Molecular Weight (g/mol) 146.233
MDL Number MFCD00008613
InChI Key PDELBHCVXBSVPJ-UHFFFAOYSA-N
Synonym 2,4,6-trimethylstyrene, 1,3,5-trimethyl-2-vinylbenzene, 2-vinylmesitylene, mesitylethylene, vinylmesitylene, styrene, 2,4,6-trimethyl, unii-89fv7yq871, benzene, 2-ethenyl-1,3,5-trimethyl, 2,6-trimethylstyrene, styrene,4,6-trimethyl
PubChem CID 13036
IUPAC Name 2-ethenyl-1,3,5-trimethylbenzene
SMILES CC1=CC(=C(C(=C1)C)C=C)C

Specifications

Color Yellow
Boiling Point 206°C
Quantity 1 mL
Formula Weight 146.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,6-Trimethylstyrene (stabilized with TBC)
TSCA No
Recommended Storage Refrigerator