accessibility menu, dialog, popup

UserName

CH3CH3CH2H3C

CAS RN 769-25-5

IUPAC Name: 2-ethenyl-1,3,5-trimethylbenzene
Synonyms: 2-ethenyl-1,3,5-trimethylbenzene
Molecular Weight (g/mol): 146.23
Molecular Formula: C11H14
InChi Key: PDELBHCVXBSVPJ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C=C)C(C)=C1
Boiling Point 
  • (2)
  • (3)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (5)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (5)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)