missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylstyrene, 95%, stab. with 500ppm 4-tert-butylcatechol

Catalog No. AAA1597403
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1597403 1 g
AAA1597406 5 g
AAA1597414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1597403 Supplier Thermo Scientific Chemicals Supplier No. A1597403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 769-25-5 | C11H14 | 146.233 g/mol

2,4,6-Trimethylstyrene is used as an intermediate to produce other chemicals like 2-Methoxy-1-(2, 4, 6-trimethyl-phenyl)-ethanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,6-Trimethylstyrene is used as an intermediate to produce other chemicals like 2-Methoxy-1-(2, 4, 6-trimethyl-phenyl)-ethanone.

Solubility
Soluble in methanol.

Notes
It is sensitive to light. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 769-25-5
Molecular Formula C11H14
Molecular Weight (g/mol) 146.233
MDL Number MFCD00008613
InChI Key PDELBHCVXBSVPJ-UHFFFAOYSA-N
Synonym 2,4,6-trimethylstyrene, 1,3,5-trimethyl-2-vinylbenzene, 2-vinylmesitylene, mesitylethylene, vinylmesitylene, styrene, 2,4,6-trimethyl, unii-89fv7yq871, benzene, 2-ethenyl-1,3,5-trimethyl, 2,6-trimethylstyrene, styrene,4,6-trimethyl
PubChem CID 13036
IUPAC Name 2-ethenyl-1,3,5-trimethylbenzene
SMILES CC1=CC(=C(C(=C1)C)C=C)C

Specifications

Density 0.908
Boiling Point 208°C to 209°C
Flash Point 75°C (167°F)
Refractive Index 1.5295
Quantity 1 g
Beilstein 1904365
Solubility Information Soluble in methanol.
Formula Weight 146.23
Percent Purity 95%
Chemical Name or Material 2,4,6-Trimethylstyrene, stabilized with 500ppm 4-tert-butylcatechol
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 212-205-4
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only