missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylpyridine 98.0+%, TCI America™

Supplier:  TCI America T0716500ML

 View more versions of this product

Catalog No. T0716500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2,4,6-Trimethylpyridine
-43°C
C8H11N
500 mL
2,4,6-collidine, gamma-collidine, s-collidine, pyridine, 2,4,6-trimethyl, collidine, sym-collidine, 2,4,6-kollidin, unii-7ie4bk5j5v, 2,4,6-trimethyl-pyridine, .gamma.-collidine
CC1=CC(C)=NC(C)=C1
121.18
121.18
Liquid
108-75-8
171°C
MFCD00006338
1992
BWZVCCNYKMEVEX-UHFFFAOYSA-N
2,4,6-trimethylpyridine
7953
≥98.0% (GC,T)

Chemical Identifiers

108-75-8
121.18
BWZVCCNYKMEVEX-UHFFFAOYSA-N
7953
CC1=CC(C)=NC(C)=C1
C8H11N
MFCD00006338
2,4,6-collidine, gamma-collidine, s-collidine, pyridine, 2,4,6-trimethyl, collidine, sym-collidine, 2,4,6-kollidin, unii-7ie4bk5j5v, 2,4,6-trimethyl-pyridine, .gamma.-collidine
2,4,6-trimethylpyridine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0715

RTECSNumber : UU0970000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions