accessibility menu, dialog, popup

UserName

CH3CH3NH3C

CAS RN 108-75-8

IUPAC Name: 2,4,6-trimethylpyridine
Synonyms: 2,4,6-collidine
Molecular Weight (g/mol): 121.18
Molecular Formula: C8H11N
InChi Key: BWZVCCNYKMEVEX-UHFFFAOYSA-N
SMILES: CC1=CC(C)=NC(C)=C1
Boiling Point 
  • (3)
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (7)
  • (3)
  • (3)
  • (4)
  • (3)
  • (7)
  • (1)
  • (3)
  • (1)
  • (2)
  • (3)
  • (7)
  • (1)
  • (1)