missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trimethylpyridine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | -43°C |
| Boiling Point | 171°C |
| Quantity | 25 mL |
| UN Number | 1992 |
| Formula Weight | 121.18 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Liquid |
| Chemical Name or Material | 2,4,6-Trimethylpyridine |
Safety and Handling
| EINECSNumber | (5)-0715 |
| RTECSNumber | UU0970000 |
| TSCA | Yes |