accessibility menu, dialog, popup

UserName

CAS RN 6095-82-5

CH3CH3NCSH3C

CAS RN 6095-82-5

IUPAC Name: 2-isothiocyanato-1,3,5-trimethylbenzene
Synonyms: 2-isothiocyanato-1,3,5-trimethylbenzene
Molecular Weight (g/mol): 177.27
Molecular Formula: C10H11NS
InChi Key: KKYMYPLVBCVDPL-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(N=C=S)C(C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results