missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trimethylbenzoic Acid 98.0+%, TCI America™
$55.84 - $128.98
Chemical Identifiers
| CAS | 480-63-7 |
|---|---|
| Molecular Formula | C10H12O2 |
| Molecular Weight (g/mol) | 164.20 |
| MDL Number | MFCD00002481 |
| InChI Key | FFFIRKXTFQCCKJ-UHFFFAOYSA-N |
| Synonym | mesitoic acid, mesitylenecarboxylic acid, beta-isodurylic acid, benzoic acid, 2,4,6-trimethyl, .beta.-isodurylic acid, unii-4pu2hg6787, 2-mesitylenecarboxylic acid, 2,4,6-trimethyl-benzoic acid, 2,6-trimethylbenzoic acid, benzoic acid,4,6-trimethyl |
| PubChem CID | 10194 |
| ChEBI | CHEBI:64830 |
| IUPAC Name | 2,4,6-trimethylbenzoic acid |
| SMILES | CC1=CC(C)=C(C(O)=O)C(C)=C1 |
Chemical Identifiers
| 480-63-7 | |
| 164.20 | |
| FFFIRKXTFQCCKJ-UHFFFAOYSA-N | |
| 10194 | |
| 2,4,6-trimethylbenzoic acid |
| C10H12O2 | |
| MFCD00002481 | |
| mesitoic acid, mesitylenecarboxylic acid, beta-isodurylic acid, benzoic acid, 2,4,6-trimethyl, .beta.-isodurylic acid, unii-4pu2hg6787, 2-mesitylenecarboxylic acid, 2,4,6-trimethyl-benzoic acid, 2,6-trimethylbenzoic acid, benzoic acid,4,6-trimethyl | |
| CHEBI:64830 | |
| CC1=CC(C)=C(C(O)=O)C(C)=C1 |
Specifications
| 480-63-7 | |
| Red-White | |
| MFCD00002481 | |
| mesitoic acid, mesitylenecarboxylic acid, beta-isodurylic acid, benzoic acid, 2,4,6-trimethyl, .beta.-isodurylic acid, unii-4pu2hg6787, 2-mesitylenecarboxylic acid, 2,4,6-trimethyl-benzoic acid, 2,6-trimethylbenzoic acid, benzoic acid,4,6-trimethyl | |
| CC1=CC(C)=C(C(O)=O)C(C)=C1 | |
| 164.20 | |
| CHEBI:64830 | |
| ≥98.0% (GC,T) | |
| 2,4,6-Trimethylbenzoic Acid |
| 154°C | |
| C10H12O2 | |
| 10 g | |
| FFFIRKXTFQCCKJ-UHFFFAOYSA-N | |
| 2,4,6-trimethylbenzoic acid | |
| 10194 | |
| 164.20 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (3)-4518
RTECSNumber : DI0887010
TSCA : Yes