accessibility menu, dialog, popup

UserName

CAS RN 480-63-7

IUPAC Name: 2,4,6-trimethylbenzoic acid
Synonyms: 2,4,6-trimethylbenzoic acid
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: FFFIRKXTFQCCKJ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(C(O)=O)C(C)=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)