missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethylbenzoic Acid 98.0+%, TCI America™

Catalog No. T142125G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T142125G 25 g
T142110G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T142125G Supplier TCI America Supplier No. T142125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 480-63-7
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00002481
InChI Key FFFIRKXTFQCCKJ-UHFFFAOYSA-N
Synonym mesitoic acid, mesitylenecarboxylic acid, beta-isodurylic acid, benzoic acid, 2,4,6-trimethyl, .beta.-isodurylic acid, unii-4pu2hg6787, 2-mesitylenecarboxylic acid, 2,4,6-trimethyl-benzoic acid, 2,6-trimethylbenzoic acid, benzoic acid,4,6-trimethyl
PubChem CID 10194
ChEBI CHEBI:64830
IUPAC Name 2,4,6-trimethylbenzoic acid
SMILES CC1=CC(C)=C(C(O)=O)C(C)=C1

Specifications

Melting Point 154°C
Color Red-White
Quantity 25 g
Formula Weight 164.20
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,6-Trimethylbenzoic Acid
EINECSNumber (3)-4518
RTECSNumber DI0887010
TSCA Yes