missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trimethylbenzoic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 154°C |
| Color | Red-White |
| Quantity | 10 g |
| Formula Weight | 164.20 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,4,6-Trimethylbenzoic Acid |
Safety and Handling
| EINECSNumber | (3)-4518 |
| RTECSNumber | DI0887010 |
| TSCA | Yes |