missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxytoluene, 97%

Catalog No. AAL0215514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0215514 25 g
AAL0215506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0215514 Supplier Thermo Scientific Chemicals Supplier No. L0215514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14107-97-2 | C10H14O3 | 182.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14107-97-2
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
MDL Number MFCD00008375
InChI Key TZPKFPYZCMHDHL-UHFFFAOYSA-N
PubChem CID 84201
IUPAC Name 1,3,5-trimethoxy-2-methylbenzene
SMILES CC1=C(C=C(C=C1OC)OC)OC

Specifications

Melting Point 27°C to 28°C
Boiling Point 100°C to 105°C (1 mmHg)
Refractive Index 1.531
Quantity 25 g
Beilstein 2048952
Synonym 2,4,6-trimethoxytoluene, benzene, 1,3,5-trimethoxy-2-methyl, acmc-1bvuf, toluene, 2,4,6-trimethoxy, 1,3,5-trimethoxy-2-methyl-benzene, #, benzene,1,3,5-trimethoxy-2-methyl
Formula Weight 182.22
Percent Purity 97%
Chemical Name or Material 2,4,6-Trimethoxytoluene
EINECSNumber 237-962-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only