accessibility menu, dialog, popup

UserName

CAS RN 14107-97-2

IUPAC Name: 1,3,5-trimethoxy-2-methylbenzene
Synonyms: 1,3,5-trimethoxy-2-methylbenzene
Molecular Weight (g/mol): 182.22
Molecular Formula: C10H14O3
InChi Key: TZPKFPYZCMHDHL-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(C)C(OC)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results