missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4,6-Trimethoxyboroxin 96.0+%, TCI America™
Supplier: TCI America T1581100G
Specifications
| 2,4,6-Trimethoxyboroxin | |
| Yellow | |
| MFCD00005946 | |
| 2924 | |
| ZFMOJHVRFMOIGF-UHFFFAOYSA-N | |
| trimethoxy-1,3,5,2,4,6-trioxatriborinane | |
| 66880 | |
| ≥96.0% (T) |
| 102-24-9 | |
| C3H9B3O6 | |
| 100 g | |
| trimethoxyboroxine, trimethoxyboroxin, boroxin, trimethoxy, trimethoxycyclotriboroxane, 2,4,6-trimethoxyboroxin, triboron trimethyl hexaoxide, boroxin, 2,4,6-trimethoxy, boric acid h3b3o6 , trimethyl ester, trimethoxy-boroxi, trimethoxyboroxole | |
| COB1OB(OC)OB(OC)O1 | |
| 173.53 | |
| 173.53 | |
| Liquid |
Chemical Identifiers
| 102-24-9 | |
| 173.53 | |
| ZFMOJHVRFMOIGF-UHFFFAOYSA-N | |
| 66880 | |
| COB1OB(OC)OB(OC)O1 |
| C3H9B3O6 | |
| MFCD00005946 | |
| trimethoxyboroxine, trimethoxyboroxin, boroxin, trimethoxy, trimethoxycyclotriboroxane, 2,4,6-trimethoxyboroxin, triboron trimethyl hexaoxide, boroxin, 2,4,6-trimethoxy, boric acid h3b3o6 , trimethyl ester, trimethoxy-boroxi, trimethoxyboroxole | |
| trimethoxy-1,3,5,2,4,6-trioxatriborinane |
Safety and Handling
EINECSNumber : (5)-3868
RTECSNumber : ED8720000
TSCA : Yes