missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxyboroxin 96.0+%, TCI America™

Catalog No. p-7136849
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T158125G 25 g
T1581100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T158125G Supplier TCI America Supplier No. T158125G
Only null left

Chemical Identifiers

CAS 102-24-9
Molecular Formula C3H9B3O6
Molecular Weight (g/mol) 173.53
MDL Number MFCD00005946
InChI Key ZFMOJHVRFMOIGF-UHFFFAOYSA-N
Synonym trimethoxyboroxine, trimethoxyboroxin, boroxin, trimethoxy, trimethoxycyclotriboroxane, 2,4,6-trimethoxyboroxin, triboron trimethyl hexaoxide, boroxin, 2,4,6-trimethoxy, boric acid h3b3o6 , trimethyl ester, trimethoxy-boroxi, trimethoxyboroxole
PubChem CID 66880
IUPAC Name trimethoxy-1,3,5,2,4,6-trioxatriborinane
SMILES COB1OB(OC)OB(OC)O1

Specifications

Color Yellow
Quantity 25 g
UN Number 2924
Formula Weight 173.53
Percent Purity ≥96.0% (T)
Physical Form Liquid
Chemical Name or Material 2,4,6-Trimethoxyboroxin
EINECSNumber (5)-3868
RTECSNumber ED8720000
TSCA Yes