missing translation for 'onlineSavingsMsg'
Learn More

2,4,6-Trimethoxybenzyl alcohol, 97%

Catalog No. AAH50348MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50348MD 250 mg
AAH5034803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50348MD Supplier Thermo Scientific Chemicals Supplier No. H50348MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 61040-78-6 | C10H14O4 | 198.218 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 61040-78-6
Molecular Formula C10H14O4
Molecular Weight (g/mol) 198.218
MDL Number MFCD00060356
InChI Key CFXXBVNHYJQNKS-UHFFFAOYSA-N
PubChem CID 602515
IUPAC Name (2,4,6-trimethoxyphenyl)methanol
SMILES COC1=CC(=C(C(=C1)OC)CO)OC

Specifications

Melting Point 60°C
Quantity 250 mg
Synonym 2,4,6-trimethoxyphenyl methanol, 2,4,6-trimethoxybenzyl alcohol, 2,4,6-trimethoxyphenyl methan-1-ol, 2,4,6-trimethoxy-benzyl alcohol, benzenemethanol,2,4,6-trimethoxy, benzenemethanol, 2,4,6-trimethoxy, 2,4,6-trimethoxy-phenyl-methanol, 2,4,6-trimethoxyphenyl methanol #, 3-amino-n-1,1-dimethylethoxy carbonyl-l-alanine methyl ester
Formula Weight 198.22
Percent Purity 97%
Chemical Name or Material 2,4,6-Trimethoxybenzyl alcohol
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only